missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ HDPE Wide Neck Square Reagent Bottles
Shop All Bürkle Products
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
750 mL
1000 mL
1500 mL
2500 mL
4000 mL
missing translation for 'unitSize'
Each
Pack of 6
Beskrivelse
- HDPE, transparent
- PP tamper-evident screw cap, blue
- UN approval for bulk goods
Tekniske data
Tekniske data
| Capacity (Metric) | 1000 mL |
| Color | Transparent |
| Diameter (Metric) Inner Neck | 43 mm |
| Material | HDPE |
| Length (Metric) | 85 mm |
| Width (Metric) | 85 mm |
| Thread Size (Metric) | 54 mm |
| Closure Color | Blue |
| Certifications/Compliance | UN Approval |
| Closure Included | Yes |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion