missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ HDPE Wide Neck Square Reagent Bottles
2.65 GBP valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Use promo code "23267" to get your promotional price.
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
750 mL
1000 mL
1500 mL
2500 mL
4000 mL
Description
- HDPE, transparent
- PP tamper-evident screw cap, blue
- UN approval for bulk goods
Specifications
Capacity (Metric) | 250 mL |
Color | Transparent |
Diameter (Metric) Outer Neck | 45 mm |
Dimensions (L x W x H) | 59 x 59 x 130 mm |
Diameter (Metric) Inner Neck | 35 mm |
For Use With (Application) | Foodstuffs |
Material | HDPE |
Neck ID (Metric) | 35 mm |
Weight (Metric) | 0.0395 kg |
Width (Metric) | 59 mm |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'