missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ S-Series Head Cover

Product Code. p-8001513
Click to view available options
Size:
Small
Large
Unit Size:
Each

Product Code. 11775314

Brand: 3M™ 7000000142

Only null left
Add to basket
Edge
Add to basket

Suitable for applications that require frequent replacement of the entire headtop assembly, such as pharmaceutical manufacturing or where there is less soiling of the outer fabric. The 3M Versaflo S-Series Head Cover is compatible with many PAPR systems.

Suitable for applications that require frequent replacement of the entire headtop assembly, such as pharmaceutical manufacturing or where there is less soiling of the outer fabric. For disposal, the suspension assembly and air inlet components can be quickly removed and separated for segregated waste streams, where facilities are available.
  • Head cover with integrated head suspension providing head and face coverage.
  • Provides eye and face protection to EN166—liquid splash and low energy impact (grade F)—the same classification for impact resistance as safety spectacles.
  • Manufactured using a general purpose, cost effective laminated film fabric.
  • Entry-level head cover for powered air respiratory protection.
  • No assembly required.
  • Ready to use out of the box.
  • Excellent field of view, particularly downwards.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 166:2:F:3,‎ EN 12941 TH3,‎ CE Approved,‎ EN14594 3A
Color White
Material Spunbound Polypropylene
Product Type Headcover
Size Large
For Use With (Application) Gas, Vapor and Particulates,‎ Liquid Splash,‎ Low Energy Flying Particles / Dust
Includes Integrated Head Suspension
Material (Lens) PETG
Weight (Metric) 110 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.