Learn More
3M™ Attest™ Rapid Readout Biological Indicators
Monitors the effectiveness of the steam sterilization process and provides results within 3 hours. 3M™ Attest™ Rapid Readout Biological Indicators provide the best assurance of sterility by challenging the sterilizer with quantifiable, highly resistant spores.
Brand:  3M™ 7000053661
Additional Details : Weight : 0.63900kg
Product Code. 11983604
  • £390.00 / Pack of 50
In Stock
to see stock.
Add to basket
Description
Description
  • Easy to use and interpret with visual color readout
  • Self-contained biological indicators significantly reduce possibility of contamination
  • Minimizes false positives
Specifications
Specifications
Autoclavable
EN886-1-2 et ISO 11138-1-3
Biological Indicator
50 Pk.