missing translation for 'onlineSavingsMsg'
Learn More

Synergy Healthcare™ Healthcare Disposal Container

Product Code. 12993475
Click to view available options
Capacity (Metric):
24 L
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12993475

Brand: Synergy Healthcare™ CW04165

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Manufactured from strong corrugated board, for use with dry aerosol and glass. Specially made for landfill disposal. Letter box type opening. Flat packed.

Specifications

Capacity (Metric) 24 L
Height (Metric) 450 mm
Quantity 10 Pk.
For Use With (Application) Collect aerosols and Glass
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.