Learn More
Gibco™ Fetal Bovine Serum, Value, heat inactivated (formerly USDA-approved in North America or qualified, Brazil in other regions)
Gibco fetal bovine sera offers an excellent value for basic cell culture, specialty research, and specific assays. It has earned the trust of researchers with consistent quality and award-winning support that helps meet research needs and budget requirements.
£46.93 - £438.00
Specifications
Content And Storage | Store at <-10°C. |
---|---|
Age | Fetal |
Species | Cattle/Bovine |
Country of Origin | Refer to CoA for specific origin |
Product Type | Fetal Bovine Serum |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||
17573595
|
Gibco™
A5209501 |
50 mL | One Shot |
£46.93
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17583595
|
Gibco™
A5209502 |
10 x 50 mL | One Shot |
£438.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17593595
|
Gibco™
A5256801 |
500 mL | Bottle |
£372.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
Gibco Serum Delivers
- ISO 13485-certified, processed in FDA-registered facilities
- Triple filtered at 0.1 μm
- Gibco bottle is easier to use in hood, reduces risk of contamination, and helps you perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
All products may not be available in all regions due to importation regulations
US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that help you meet your research needs and budget requirements.
- Sera category: Value
- Sera application: Cell culture with robust cell lines, good value for basic research
- Testing: Up to 30 quality tests
- Endotoxin level: ≤20 EU/mL (levels routinely ≤10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
- Origin: Refer to CoA for specific origin
Spezifikation
Store at <-10°C. | |
Cattle/Bovine | |
Fetal Bovine Serum | |
Sterile | |
Value | |
≤20 EU/mL | |
Yes | |
Value | |
USDA Safety Testing Performed for Mexican and Australian origins |
Fetal | |
Refer to CoA for specific origin | |
Heat-inactivated | |
Triple-filtered, 0.1 μm | |
Up to 30 quality tests including 9CFR virus testing* endotoxin, performance (*Please note: If manufactured in the UK, Full 9CFR except for Rabies and BlueTongue Virus which is tested via EMA) | |
Less waste | |
≤25 mg/dL | |
Dry Ice |
Sicherheit und Handhabung
missing translation for 'shelfLife' : 4 Years
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.