missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keystone Cleanroom Products 1073B Tyvek Autoclave Pouch, with Steam Indicator Dot
Tyvek Pouch that offers high breathability while providing a barrier from microbial contamination after sterilization.
£268.00 - £1390.00
Specifications
Color | White |
---|---|
For Use With (Equipment) | Pharmaceutical Equipment |
Material | Tyvek |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
17748327
|
Keystone Cleanroom Products
COVMGTVKUB12X16 |
40.6 x 30.5 cm (16 x 12 in.) |
£481.00
Pack of 100 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17788357
|
Keystone Cleanroom Products
COVMGTVKUB24X36 |
61 x 91.4 cm (24 x 36 in.) |
£530.00
Pack of 50 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17758337
|
Keystone Cleanroom Products
COVMGTVKUB15X18 |
45.7 x 38.1 cm (18 x 15 in.) |
£670.00
Pack of 100 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17718357
|
Keystone Cleanroom Products
COVMGTVKUB1X14 |
35.6 x 2.54 cm (14 x 1 in.) |
£307.00
Pack of 100 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17788367
|
Keystone Cleanroom Products
COVMGTVKUB30X30 |
76.2 x 76.2 cm (30 x 30 in.) |
£545.00
Pack of 50 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17708387
|
Keystone Cleanroom Products
COVMGTVKUB36X36 |
91.4 x 91.4 cm (36 x 36 in.) |
£600.00
Pack of 50 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17728387
|
Keystone Cleanroom Products
COVMGTVKUB36X42 |
106.7 x 91.4 cm (42 x 36 in.) |
£650.00
Pack of 50 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17748387
|
Keystone Cleanroom Products
COVMGTVKUB36X48 |
121.9 x 91.4 cm (48 x 36 in.) |
£860.00
Pack of 50 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17768387
|
Keystone Cleanroom Products
COVMGTVKUB36X60 |
152.4 x 91.4 cm (60 x 36 in.) |
£960.00
Pack of 50 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17718397
|
Keystone Cleanroom Products
COVMGTVKUB3X6 |
15.2 x 7.6 cm (6 x 3 in.) |
£268.00
Pack of 100 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
17841081
|
Keystone Cleanroom Products
COVMGTVKUB57X57 |
145 x 145 cm (57 x 57 in.) |
£1390.00
Pack of 50 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
17746899
|
Keystone Cleanroom Products
COVMGTVKUB32X32 |
81 x 81 cm (32 x 32 in.) |
£550.00
Pack of 50 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
Description
- All Tyvek construction for strength and microbial barrier
- Designed for steam sterilization
- Strong Factory seals
- Heat Seal Closure
- Recommended for sterilization of pharmaceutical equipment and Components
- Manufactured and packaged in a ISO 6 cleanroom
- Improves the efficiency of the Parts Preparation Department
Specifications
White | |
Pharmaceutical Equipment | |
Tyvek |