Learn More
Eppendorf™ Heat Sealer S200
Product Code. p-7159215 Shop All Eppendorf Products
Click to view available options
Plug Type:
GB Plug
Unit Size:
Each
Product Code. 15785665
Brand: Eppendorf™ 5392000030
Provides user-friendly and reliable hermetic sealing of a variety of multiwell plates.
  • Eliminates evaporation in PCR
  • Adjustable sealing temperatures for more flexibility
  • Seals 96- and 384-well plates
  • Sealing time selectable from 1 to 9 seconds in 0.5 second increments
  • Integrated thermostat prevents overheating
  • Powerful handle for ergonomic operation
  • PCR plate adapter included
TRUSTED_SUSTAINABILITY
Specifications
Hertz 50/60 Hz
Voltage 100/230 V
Dimensions (L x W x H) 220 × 321 × 425 mm
Electrical Requirements 100/230 V, 50/60 Hz
For Use With (Equipment) 96- and 384-well plates
Plug Type GB Plug
Sealing Temperature Range 125°C to 200°C in 1°C increments