missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fistreem International™ Heating Element, Loughborough Glass Water Still
Suitable replacement heating element for the reliable and robust Loughborough Glass Water Still. Fistreem International™ Heating Element, Loughborough Glass Water Still is heavy duty and chromium-plated.
Brand: Fistreem International™ WH/A/220
This item is not returnable.
View return policy
Description
- Heavy duty, 3kW, chromium plated heating element.
- Thermostatically protected.
Specifications
200 V | |
3000 w | |
WGS-560/570 4.0 L and 8.0 L Stills | |
Heating Element |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction