missing translation for 'onlineSavingsMsg'
Learn More

Heidolph instruments Heating Baths

This aluminum bath for superior heat transfer comes with two insulated safety handles and a safety rim for secure mounting on the hot plate.

Brand:  Heidolph instruments 504-93000-00

 View more versions of this product

Product Code. 10059090

  • £336.00 / Each
Estimated Shipment: 11-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 1-l Heating bath for oil
  • Oil bath Heidolph for oil up to 250°C 
  • Aluminum bath for superior heat transfer
  • Comes with two insulated safety handles and safety rim for secure mounting on the hot plate
  • Maximum temperature of 250°C
  • Accessory for magnetic stirrers
Specifications

Specifications

1 L
Heating Bath for Oil, 1 L
Oil
Heating Bath
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers