Learn More
Axygen™ Heating Block
Interchangeable aluminum block to accommodate PCR plates and a variety of common life science vessels.
Brand: Axygen™ I-4010-D
This item is not returnable.
View return policy
Description
- No additional tools are required for block exchanges
- The blocks utilize a magnetic adhesion technology and are self-recognized once exchanged
- The combination of heating, cooling, and mixing with a versatile block selection makes the Axygen Microtube Shaker ideal for many life science laboratories in the segments of molecular biology, biochemistry, and clinical chemistry
- Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
Specifications
5 mL | |
0.3 kg | |
Flat Bottom | |
8 |