Learn More
Axygen™ Heating Block
Interchangeable aluminum block to accommodate PCR plates and a variety of common life science vessels.
Brand:  Axygen™ I-4010-F
 View more versions of this product
Product Code. 17842617
  • £554.71 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • No additional tools are required for block exchanges
  • The blocks utilize a magnetic adhesion technology and are self-recognized once exchanged
  • The combination of heating, cooling, and mixing with a versatile block selection makes the Axygen Microtube Shaker ideal for many life science laboratories in the segments of molecular biology, biochemistry, and clinical chemistry
  • Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15 mL
0.3 kg
Flat Bottom
8