Learn More
Cole-Parmer™ Heating Block for Electrothermal™ RS-250D-S Shaking Reaction Stations
Product Code. 11791467
Click to view available options
Holds:
4 x 96-well PTFE Microplates
40 x 24 mm Tubes
96 x 16 mm Tubes
Accessory heating blocks to keep your reactions running smoothly.
  • Compatible with Electrothermal RS9000 Reaction Stations
One year warranty
  • Specifications
    Description Heating Block for Reaction System
    For Use With (Equipment) RS-250D-S Shaking Reaction Stations
    Holds 96 x 16 mm Tubes
    No. of Places 96
    Type Heating Block
    Warranty 1 Year
    Diameter (English) Outside 0.6 in.
    Diameter (Metric) Outside 16 mm