missing translation for 'onlineSavingsMsg'
Learn More

JP Selecta™ Heating Tape

POA

Specifications

Width (Metric) 25mm
Product Type Heating Tape
Temperature (Metric) 210°C
For Use With (Application) For heating small areas such as heating ducts, distillation, valves and pipes
View More Specs

Description

Description

Specifications

Specifications

25mm
Heating Tape
210°C
For heating small areas such as heating ducts, distillation, valves and pipes
Videos
SDS
Documents

Documents

Product Certifications