Learn More
3M™ Heavy Duty QRS Breathing Tube
3M™ Heavy Duty QRS Breathing Tube for 3M™ Adflo™ Powered Air Respirator and 3M™ Versaflo™ Supplied Air Regulator V-500E.
Brand: 3M™ 7000044527
57.45 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 7.04000kg
Description
- The new welding helmets and faceshields from 3M feature the QRS type of breathing tube (Quick Release Swivel), which enables one-handed connection and release
- The protection class of Speedglas welding helmet 9100FX Air is reduced to TH2 when the adapter is used
- You can extend the service life of the breathing tube by using a tube cover
Specifications
Heavy Duty Rubber |