missing translation for 'onlineSavingsMsg'
Learn More

Heavy Wall Rimless Borosilicate Glass Test Tube

Brand:  Samco G033/01

Product Code. 13028403

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

ISO 4142:2002
Borosilicate Glass
100 Pack
10 mm
75 mm
100
No
Test Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heavy Wall Rimless Borosilicate Glass Test Tube > Diameter Outer: 10mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.