Learn More
Heidolph™ Hei-FLOW Core 120 Multi
Peristaltic pump ideal for the simple conveying of all media.
Brand: Heidolph™ 523-50013-02
This item is not returnable.
View return policy
Description
- The pump drive of this model makes it possible to switch between a single channel and multichannel pump head. By simply flipping the switch, it can also be conveyed in an anti-clockwise direction.
- Depending on the adapted pump head, a flow rate of 0.83 to 861 mL/min for the single-channel and 0.005 to 364 mL/min for the multichannel pump head is guaranteed. The Value 01 conveys with a control accuracy of ± 5% at a speed range of 10 to 120 rpm.
Specifications
UK Plug | |
Analog | |
230 V |
Digital Variable Speed | |
IP55 |