Learn More
Heidolph™ Hei-Mix L
The Hei-Mix L was especially developed for applications with thermolabile substances.
Brand:  Heidolph™ 505-00000-02
Product Code. 17303000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Stirring up to 20 liters without heating
  • The Hei-Mix L was especially developed for applications with thermolabile substances. This is because this model has no heating, and the heat from the motor is also not transferred to the footprint.
  • The Hei-Mix L magnetic stirrer without heating function is ideal for applications up to 20 liters and can gently stir your solution in a speed range of 100 to 1400 rpm. In addition, the rotation speed ramps up slowly until it has reached the desired value. This prevents splashes, uneven starts, or jumps of the stirring rod.
  • The patented Kera-Disk™ footprint made of aluminum with a ceramic coating, is chemical-resistant, scratch-proof, and has a diameter of 145 mm. This unique size makes it suitable for use with many accessories and samples on a small footprint.
Specifications
Specifications
1
230 V
UK