Learn More
Heidolph™ Hei-Mix S
The Hei-Mix S is the first choice for titrations and biochemical applications such as those associated with operating a small fermenter.
Brand: Heidolph™ 503-02000-13
282.15 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 1.10000kg
Description
- Stirring up to 5 liters without heating
- The Hei-Mix S is the first choice for titrations and biochemical applications such as those associated with operating a small fermenter. The PVDF (thermoplastic fluoropolymer) footprint is of white color: In this way, color changes during the application can be precisely tracked.
- The Hei-Mix S also has a robust housing made of polyamide and is suitable for continuous operation with stirring volumes of up to 5 liters. Thanks to a wide speed range of up to 2,200 rpm, you can mix your sample quickly but gently. Efficient work is therefore guaranteed.
- In addition, you save space in your laboratory because the Hei-Mix S requires only 126 x 80 x 140 mm of space with a footprint of 104 mm in diameter.
Specifications
1 | |
230 V | |
UK |