Learn More
Heidolph™ Hei-PLATE Silver 1 Package
Package includes Hei-Tec, Pt1000 temperature sensor, and Pt1000 clamping system.
Brand: Heidolph™ 505-30080-03
This item is not returnable.
View return policy
Description
- Magnetic Stirrer Hei-Tec
- Temperature sensor Pt1000 (AISI 316Ti)
- Pt1000 clamping system (includes support rod and attachment with cable inlet)
Specifications
Aluminum Alloy | |
Magnetic Stirring Hotplate | |
UK-Style | |
Digital |
230 V | |
Round | |
Multiple |