Learn More
Helios Italquartz Quartz Crucibles
Excellent for storage of samples in the laboratory. Helios Italquartz™ Quartz Crucibles offer good thermal shock resistance up to 1000°C.
Brand:  Helios Italquartz 63P00016
 View more versions of this product
Product Code. 11704066
  • £169.00 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Chemically inert to acids (except HF)
  • Available in a wide range of volumes
  • Available in transparent or translucent color
  • Available in tall or short form
  • Resistant to corrosion
Specifications
Specifications
Transparent
Quartz
190 mL
80mm
Tall form
1000°C
75mm
Crucible
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products