Learn More
Helios Italquartz Quartz Crucibles
Excellent for storage of samples in the laboratory. Helios Italquartz™ Quartz Crucibles offer good thermal shock resistance up to 1000°C.
Brand: Helios Italquartz 63F00508
Description
- Chemically inert to acids (except HF)
- Available in a wide range of volumes
- Available in transparent or translucent color
- Available in tall or short form
- Resistant to corrosion
Specifications
Translucent | |
Quartz | |
25 mL | |
28mm |
Short form | |
1000°C | |
47mm | |
Crucible |