Learn More
Helios Italquartz Quartz Crucibles
Excellent for storage of samples in the laboratory. Helios Italquartz™ Quartz Crucibles offer good thermal shock resistance up to 1000°C.
Brand:  Helios Italquartz 63F00515
 View more versions of this product
Product Code. 11734906
  • £152.00 / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Chemically inert to acids (except HF)
  • Available in a wide range of volumes
  • Available in transparent or translucent color
  • Available in tall or short form
  • Resistant to corrosion
Specifications
Specifications
Translucent
Quartz
130 mL
50mm
Short form
1000°C
80mm
Crucible