Learn More
3M™ Helmet Back Plate Adapter, Z3G
Z3G adapters are the slot fitting back plates with 25 mm wide (G fitting) tongue. It is used to mount Peltor™ Multi-Visors and Ear Muffs fitted with the P3 mounts to 3M™ Safety Helmets or selected other manufacturers helmets.
Brand: 3M™ 4100010854
Code : NEW
Additional Details : Weight : 11.85300kg
Description
- Adapter to connect visors and multivisors to Safety Helmets for added face protection
- Snaps easily into most safety helmets with universal accessory slots to stop accessories becoming dislodged in use
- Z3G adaptor for mounting visors or earmuffs
- Snaps easily into the helmet universal accessory slot to stop these accessories becoming dislodged in use
Specifications
25 mm |