missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Helmet Back Plate Adaptor
3M™ Helmet Back Plate Adaptor is used to mount PELTOR Multi-Visors and PELTOR Ear Muffs fitted with the P3 mounts to 3M™ Safety Helmets.
£28.35 - £63.85
Specifications
Color | Black |
---|---|
For Use With (Equipment) | Visors, Multivisors, and Protective Helmets |
For Use With (Application) | Connecting visors to helmets |
Product Type | Helmet Back Plate Adapter |
Width (Metric) | 25 mm |
Description
- Adaptor to connect 3M™ visors and multivisors to Safety Helmets for added face protection
- Snaps easily into most safety helmets with universal accessory slots to help provide face protection
- Adaptor for mounting visors or earmuffs
- Snaps easily into the helmet's universal accessory socket to prevent these accessories from being damaged during use
Specifications
Black | |
Connecting visors to helmets | |
25 mm |
Visors, Multivisors, and Protective Helmets | |
Helmet Back Plate Adapter |