Learn More
3M™ Helmet Integrated Safety Glasses
189.00 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Lens Tint:
Clear
Gray
Yellow
Description
- Safety Glasses that easily attach to 3M™ Hard Hats for eye protection
- The integrated eyewear fit closely to the wearer's cheeks, providing excellent protection from particles, dust and approved to EN166
- Durable lens coating helps minimize physical and chemical damage
- Easy to adjust for a good fit
Specifications
Certifications/Compliance | EN 166, CE Approved |
Lens Tint | Gray |
Product Type | Safety Glasses |
For Use With (Application) | Construction and forestry |
Lens Material | Polycarbonate |