missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Hematocrit Tubes

Product Code. p-8013428
Click to view available options
Capacity (Metric):
60 μL
75 μL
Unit Size:
Pack of 5000
This item is not returnable. View return policy

Product Code. 30090386

Brand: Hirschmann™ 9100160

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Excellent for centrifuging blood in the laboratory. Hirschmann Instruments™ Hematocrit Tubes are manufactured from disposable capillary glass.

  • Not heparinized
  • Color-coded blue
  • Acc. to DIN EN ISO 12772, CE acc. IVD 98/79 EC

Specifications

Product Type Hematocrit Tube
Capacity (Metric) 60 μL
Diameter (Metric) Inner 0.9/1.0 mm
For Use With (Application) For centrifuging blood samples, not heparinized
Length (Metric) 75 ±0.5 mm
Certifications/Compliance DIN EN ISO 12772, CE, IVD 98/79 EC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.