missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Hematology Phosphate Buffer

Combine Epredia™ Phosphate Buffer with Giemsa, Wright, and Wright-Giemsa stains for excellent results.

£94.25 - £168.00

Specifications

Product Type Phosphate Buffered Saline (PBS)
View More Specs

Products 2
Product Code Brand pH Quantity Price Quantity & Availability  
Product Code Brand pH Quantity Price Quantity & Availability  
12520037
View Documents
Epredia™
89032
6.6 9.25 g N/A N/A N/A
12530037
View Documents
Epredia™
89033
7.2 12 x 9.25g N/A N/A N/A
Description

Description

Supplied as 9.25g powder.
Specifications

Specifications

Phosphate Buffered Saline (PBS)
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H371-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Hematology Phosphate Buffer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.