missing translation for 'onlineSavingsMsg'
Learn More

Hematoxylin Trihydrate, Fisher BioReagents

£39.53 - £202.00

Chemical Identifiers

CAS 517-28-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00078111
InChI Key WZUVPPKBWHMQCE-WKTCHCBJNA-N
Synonym Natural Black I
PubChem CID 45029742
SMILES [H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10012694
View Documents
Fisher BioReagents
BP2523-5
5 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10072594
View Documents
Fisher BioReagents
BP2523-100
100 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Hematoxylin Trihydrate is used mainly as a stain in microscopy.
Specifications

Chemical Identifiers

517-28-2
302.28
WZUVPPKBWHMQCE-WKTCHCBJNA-N
45029742
C16H14O6
MFCD00078111
Natural Black I
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3

Specifications

100°C
C16H14O6
15, 4672
Pass Test
[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
302.28
290 to 292nm
Report %
Powder
517-28-2
MFCD00078111
Natural Black I
WZUVPPKBWHMQCE-WKTCHCBJNA-N
(1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
45029742
356.4
Amber
Hematoxylin Trihydrate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS
Documents

Documents

Product Certifications