missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hemoglobin epsilon Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12361-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can be used to label and isolate fetal cells from maternal blood and may be useful in prenatal diagnosis.
The epsilon globin gene (HBE) is normally expressed in the embryonic yolk sac: two epsilon chains together with two zeta chains (an alpha-like globin) constitute the embryonic hemoglobin Hb Gower I; two epsilon chains together with two alpha chains form the embryonic Hb Gower II. Both of these embryonic hemoglobins are normally supplanted by fetal, and later, adult hemoglobin. The five beta-like globin genes are found within a 45 kb cluster on chromosome 11 in the following order: 5′-epsilon - G-gamma - A-gamma - delta - beta-3′Specifications
Hemoglobin epsilon | |
Polyclonal | |
Unconjugated | |
HBE1 | |
Epsilon globin, HBE, HBE1, Hemoglobin epsilon chain, Hemoglobin subunit epsilon, hemoglobin, epsilon 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110337495, 293267, 3046 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02100 | |
Hbe1 | |
Hemoglobin epsilon Fusion Protein Ag3026 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hemoglobin epsilon Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction