missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ HEPA Air Filter

This HEPA Air Filter provides a fresh air supply for BINDER™ FD 720 Heating and Drying Chambers. Filter classification H14.

Brand:  BINDER™ 8012-1099

Additional Details : Weight : 5.85000kg

Product Code. 16454069

  • £1460.00 / Each
Estimated Shipment: 16-09-2024
to see stock.
Add to basket

Description

Description

  • Provides a fresh air supply
  • Filters greater than 99.995% of particulate matter as small as 0.3 μm
  • For use with BINDER™ FD 720 Heating and Drying Chambers
Specifications

Specifications

EN 1822-1:2009
BINDER Chambers FD 720 Drying and Heating Chambers with Forced Convection
Providing Fresh Air Supply for Unit
HEPA air filter - On fresh air supply for unit; filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% (at 0.3 µm))
Air Filter
HEPA (Filter Classification H14)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers