missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ HEPA Air Filter for Series FP Avantgarde.Line Drying and Heating Chambers

Fresh air supply for the Series FP Avantgarde.Line Drying and Heating Chambers.

Brand:  BINDER™ 8012-2219

Additional Details : Weight : 0.50000kg

Product Code. 17824451

  • £1460.00 / Each
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

  • Accessory for Series FP Avantgarde.Line Drying and Heating Chambers
Specifications

Specifications

EN 1822-1:2009
Removes >99.995% (at 0.3 μm)
Air Filter
H14
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers