Learn More
Cytiva (formerly Pall Lab) HEPA Capsule with Versapor™ Membrane
Capsule ensures high efficiency particulate-free air (HEPA).
Brand:  Cytiva (formerly Pall Lab) 12144
Additional Details : Weight : 0.20000kg
Product Code. 17104231
  • £247.00 / Each
Estimated Shipment: 30-08-2024
to see stock.
Description
Description
  • Porous membrane delivers high air flow rates at low differential pressure and extends filter life
  • Ensures particulate-free air 99.97% retention of 0.3 μm DOP aerosol
  • Designed for laboratory applications only
Specifications
Specifications
Acrylic Copolymer on a Non-woven Support
Sterile
3/8 in. FNPT and Hose Barb
1.2 μm
860 cm2