missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HEPA Filter

For Use With: For Water Jacket models, Steri-cycle and direct heat models (31X, 32X, 37X & 38X) and 8000 Series Incubators

Brand:  Thermo Scientific™ 760209

Code : NEW

Additional Details : Weight : 0.74700kg

Product Code. 15368111

  • £316.00 / Pack of 4
In Stock
to see stock.
Add to basket

Description

Description

Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters.

HEPA Filters

  • 99.97% filtering at 0.3μm
  • Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Specifications

Specifications

Main HEPA Filter, Value pack (4/pk)
For Water Jacket models, Steri-cycle and direct heat models (31X, 32X, 37X & 38X) and 8000 Series incubators
LED
HEPA Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers