Learn More
Heparin Cofactor II Recombinant Rabbit Monoclonal Antibody (25), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA529795
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse SerpinD1/HCII.
The product encoded by this gene is a serine proteinase inhibitor which rapidly inhibits thrombin in the presence of dermatan sulfate or heparin. The gene contains five exons and four introns. This protein shares homology with antithrombin III and other members of the alpha 1-antitrypsin superfamily. Mutations in this gene are associated with heparin cofactor II deficiency.Specifications
Heparin Cofactor II | |
Recombinant Monoclonal | |
Unconjugated | |
Serpind1 | |
AA985900; AI303446; D22S673; HC2; Hcf2; HCII; HC-II; Heparin cofactor 2; heparin cofactor II; HLS2; leuserpin 2; leuserpin-2; LS2; Protease inhibitor leuserpin-2; rls2var1; serine (or cysteine) peptidase inhibitor, clade D, member 1; serine (or cysteine) proteinase inhibitor, clade D (heparin cofactor), member 1; serine (or cysteine) proteinase inhibitor, clade D, member 1; serpin D1; serpin family D member 1; serpin peptidase inhibitor, clade D (heparin cofactor), member 1; Serpind1; THPH10 | |
Rabbit | |
Protein A | |
RUO | |
15160 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
25 | |
PBS with no preservative | |
P49182 | |
Serpind1 | |
Recombinant Mouse SerpinD1 protein (Met1-Ser478) | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.