missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Heparin Sepharose™ 6 Fast Flow Affinity Chromatography Media

USe for  separations of biomolecules with an affinity for heparin, including antithrombin III, coagulation factors and other plasma proteins.  Cytiva Life Sciences™ Heparin Sepharose™ 6 Fast Flow Affinity Chromatography Media is fast and reliable and highly suitable for process-scale purification processes.

Brand:  Cytiva 17-0998-01

Code : GE

Additional Details : CAS Number : 64-17-5 Weight : 1.07050kg


Product Code. 10526730

  • £564.00 / 50mL
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

Description

Description

  • Composed of cross-linked 6% agarose beads modified sodium heparin
  • Immobilization performed using chemically stable and oriented coupling giving high binding capacity
  • Enhanced binding capacity for antithrombin III due to oriented coupling of the ligand
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

Specifications

Specifications

50 mL
Stable in commonly used aqueous Buffers: 0.1 M NaOH, 0.05 M Sodium Acetate, 8 M Urea, 6 M Guanidine Hydrochloride, non-ionic detergents, 10% Glycerol
Heparin Sepharose 6 Fast Flow Media
45 to 165 μm
25 cm
6% Cross-Linked Agarose
20% Ethanol, 0.05 M Sodium Acetate
For Fast and Reliable Separations of Biomolecules with an Affinity for Heparin, Including Antithrombin III, Coagulation Factors and Other Plasma Proteins Highly Suitable for Process-Scale PurifiCatIon Processes
4°C to 30°C
250 to 400 cm/Hoμr
9.84 in.
100 kPa
4 to 12
Porcine heparin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Heparin Sepharose 6 Fast Flow, 50 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers