Learn More
Karl Hecht™ Heparinised Micro Hematocrit Capillaries
A fire-polished hematocrit capillary with sodium heparin.
Brand:  Karl Hecht™ 40565010
Product Code. 10460742
  • £7.10 / Pack of 100
Explore more special offers
Description
Description
  • Disposable
  • With mark at 60mm
  • With red color code
  • Fire-polished at color-coded end
  • Total capacity of approx. 18μL
  • Capacity up to mark approx. 14μL
  • With CE marking
  • 100 pieces/tube; 10 tubes/box
Specifications
Specifications
18 μL, 14 μL to mark
100 Pieces/Tube; 10 Tubes/Box
CE
0.55 mm
75 mm