Learn More
Hepatocyte Growth Factor Ab-2 Mouse Monoclonal Antibody, Epredia™
Ideal for Western Blotting, Epredia™ Hepatocyte Growth Factor Ab-2, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-775-P1ABX
337.04 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
Description
Hepatocyte Growth Factor (HGF), also known as Scatter Factor (SF) or Hepatopoietin A. Membrane receptor for HGF/SF is the product of the proto-oncogene c-met, a tyrosine kinase. HGF/SF is a pleiotropic growth factor which is produced by mesodermally derived cells, such as Kupffer cells/macrophages, endothelial cells, or hepatic fat-storing cells.Host Species: Mouse
Clone: SBF5
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human HGF protein
Molecular Weight: ∽82kDa
Recommended for:
- Western Blotting (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Hepatocyte Growth Factor Ab-2 | |
SBF5 | |
Unconjugated | |
Recombinant human HGF protein | |
RUO | |
Primary | |
Concentrated |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Growth Factor | |
Human | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.