missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ HEPES, 1M Buffer Solution
HEPES, 1M Buffer Solution
£21.58 - £1114.00
Specifications
Absorbance | UV light |
---|---|
Chemical Name or Material | HEPES buffer |
Certificate of Analysis | FDA, ISO 13485 standards |
Concentration | 1 M |
For Use With (Application) | Mammalian cell culture |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11550496
|
Gibco™
15630049 |
20 mL |
£21.58
20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11570496
|
Gibco™
15630122 |
20 x 100 mL |
£1114.00
Pack of 20 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gibco™ HEPES (N-2-Hydroxyethylpiperazine-N-2-Ethane Sulfonic Acid) is a zwitterionic organic chemical buffering agent commonly used in cell culture media. The addition of 10 – 25 mM HEPES provides extra buffering capacity when cell culture requires extended periods of manipulation outside of a CO2 incubator. HEPES is a good buffering choice for many cell culture systems because it is membrane impermeable, has limited effect on biochemical reactions, is chemically and enzymatically stable, and has very low visible and UV light absorbance.Product Use
For in vitro diagnostic use.
Dual-site cGMP Manufacturing and Quality System
Gibco™ HEPES is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ HEPES product made in our Grand Island facility (15630-106). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
UV light | |
FDA, ISO 13485 standards | |
Mammalian cell culture | |
Liquid |
HEPES buffer | |
1 M | |
7.2 to 7.5 | |
1 Each |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title