missing translation for 'onlineSavingsMsg'
Learn More

HEPES, >98%, MP Biomedicals™

Product Code. 11400832
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
250 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym 4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N?-(2-ethanesulfonic acid) sodium salt
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Color White
Physical Form Powder
Quantity 10 g
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
Formula Weight 238.3
Percent Purity >98%
Chemical Name or Material HEPES
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'