missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES, >98%, MP Biomedicals™
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
250 g
500 g
1 kg
Specifications
Specifications
Color | White |
Physical Form | Powder |
Quantity | 50 g |
IUPAC Name | 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid |
Formula Weight | 238.3 |
Percent Purity | >98% |
Chemical Name or Material | HEPES |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'