missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES, >98%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
250 g
500 g
1 kg
Unit Size:
100g
10g
1kg
250g
25g
500g
50g
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 250 g |
| IUPAC Name | 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid |
| Formula Weight | 238.3 |
| Percent Purity | >98% |
| Chemical Name or Material | HEPES |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction