missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals HEPES, 99%, for biochemistry
£40.30 - £2108.49
Chemical Identifiers
CAS | 7365-45-9 |
---|---|
Molecular Formula | C8H18N2O4S |
Molecular Weight (g/mol) | 238.30 |
MDL Number | MFCD00006158 |
InChI Key | JKMHFZQWWAIEOD-UHFFFAOYSA-N |
Synonym | 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid |
PubChem CID | 23831 |
ChEBI | CHEBI:42334 |
SMILES | OCCN1CCN(CCS(O)(=O)=O)CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10521241
|
Thermo Scientific Chemicals
172570250 |
25 g | Glass bottle |
£40.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10666572
|
Thermo Scientific Chemicals
172571000 |
100 g | Glass bottle |
£100.89
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10246130
|
Thermo Scientific Chemicals
172572500 |
250 g | Glass bottle |
£205.65
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10123742
|
Thermo Scientific Chemicals
172570010 |
1 kg | Glass bottle |
£532.65
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11936521
|
Thermo Scientific Chemicals
172570050 |
5 kg | Plastic drum |
£2108.49
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
7365-45-9 | |
238.30 | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
23831 | |
OCCN1CCN(CCS(O)(=O)=O)CC1 |
C8H18N2O4S | |
MFCD00006158 | |
2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid | |
CHEBI:42334 |
Specifications
0.08 max. at 260nm, 0.05 max. at 280nm, 0.08 max. at 250nm, (5% in water) 1 cm cell, (5% in water) 1 cm cell at 0nm | |
7365-45-9 | |
(5% in water) clear, colorless | |
Liquid or Solid | |
Authentic | |
C8H18N2O4S | |
23, V,2, 379 | |
2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid | |
23831 | |
238.3 | |
Biochemical |
(5% aq. soln.) (1cm cell) | |
234.0°C to 238.0°C | |
238°C | |
99% | |
1 % max. (1 g, 105°C, 3 hrs) | |
MFCD00006158 | |
15, 4689 | |
Solubility in water: 704g/L (20°C). | |
OCCN1CCN(CCS(O)(=O)=O)CC1 | |
238.30 | |
CHEBI:42334 | |
99% | |
HEPES |
Safety and Handling
Product Identifier
- HEPES
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific Chemicals HEPES, 99%, for biochemistry