missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES, ≥99% (Dry Basis), Ultrapure, Thermo Scientific Chemicals
348.65 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Specifications
Melting Point | ∼236°C (decomposition) |
Physical Form | Crystalline Powder |
Quantity | 1 kg |
Packaging | Plastic bottle |
Beilstein | 883043 |
Formula Weight | 238.3 |
Percent Purity | ≥99% |
Assay | Dry basis |
Chemical Name or Material | HEPES |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'