missing translation for 'onlineSavingsMsg'
Learn More

HEPES Buffer, 1M Solution, Cell Culture Grade, MP Biomedicals

Product Code. 11464805
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

1M solution pH 7.2 to 7.4 at 37°C Storage temperature: 15-30°C

TRUSTED_SUSTAINABILITY

Specifications

CAS 7365-45-9
Concentration 1 M
Physical Form Solution
Quantity 100 mL
Molecular Formula C8H18N2O4S
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular Weight (g/mol) 238.30
PubChem CID 23831
ChEBI CHEBI:42334
Grade Cell Culture
Chemical Name or Material Hepes Buffer
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'