missing translation for 'onlineSavingsMsg'
Learn More

HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™

Product Code. p-26398
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Unit Size:
1 L
100mL
500mL
This item is not returnable. View return policy

Product Code. 10041703

Brand: Fisher BioReagents BP299100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Commonly used buffering agent

HEPES is commonly used as a buffering agent.
TRUSTED_SUSTAINABILITY

Specifications

Absorbance ≤0.01(0.1M solution) at 260nm, 0.004 max. (0.1M solution) at 280nm
Chemical Name or Material HEPES Buffer
Name Note 1M Solution, pH 7.3
CAS 7365-45-9, 7732-18-5
Color Undesignated
Concentration 0.95 to 1.05 M
pH 7.2 to 7.5
Physical Form Liquid
Quantity 100 mL
DNase DNase free
Purity Grade Notes DNase-, RNase- and Protease-Free
Identification Pass Test
Molecular Formula C8H18N2O4S
Packaging Poly Bottle
Protease Protease free
Merck Index 15, 4689
Synonym N-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid
Formula Weight 238.197
Grade Molecular Biology
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.