missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES-buffered saline, pH 7.0 (5X)

£65.22 - £118.59

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15463469
View Documents
Thermo Scientific Chemicals
J60753.AK
250 mL
£65.22
250mL
Please to purchase this item. Need a web account? Register with us today!
15473469
View Documents
Thermo Scientific Chemicals
J60753.AP
500 mL
£118.59
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HEPES is widely used in cell culture. HEPES is used to neutralize the complex proteins in growth medium that may inactivate trypsin during trypsinization. N-2-hydroxyethylpiperazine-N-2?-ethanesulfonic acid (Hepes) has been tested as an alternative to tris-(hydroxymethyl)aminoethane (Tris) for hydrogen-ion buffering of algal cultures. Separation of human platelets from plasma proteins including factor VIII VWF by a combined albumin gradient-gel filtration method by using HEPES buffer. May be used in isoelectric focusing applications and as a buffer in the quantitative and selective measurement of antigen-antibody reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is widely used in cell culture. HEPES is used to neutralize the complex proteins in growth medium that may inactivate trypsin during trypsinization. N-2-hydroxyethylpiperazine-N-2′-ethanesulfonic acid (Hepes) has been tested as an alternative to tris-(hydroxymethyl)aminoethane (Tris) for hydrogen-ion buffering of algal cultures. Separation of human platelets from plasma proteins including factor VIII VWF by a combined albumin gradient-gel filtration method by using HEPES buffer. May be used in isoelectric focusing applications and as a buffer in the quantitative and selective measurement of antigen-antibody reactions.

Solubility
Soluble in H2O: 1 M at 20°C, clear, colorless

Notes
Contains 125mM HEPES and 750mM sodium chloride, at pH 7.0
Specifications

Chemical Identifiers

7365-45-9
238.30
JKMHFZQWWAIEOD-UHFFFAOYSA-N
CHEBI:42334
OCCN1CCN(CCS(O)(=O)=O)CC1
C8H18N2O4S
MFCD00006158
23831
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid

Specifications

7365-45-9
7.0
C8H18N2O4S
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
23831
HEPES-buffered saline
5X
Liquid
Odorless
Soluble in H2O: 1 M at 20°C,clear,colorless
OCCN1CCN(CCS(O)(=O)=O)CC1
238.30
CHEBI:42334
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only