missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
Commonly used buffering agent.
Brand: Fisher BioReagents BP310-100
64.65 GBP valid until 2024-12-31
Use promo code "21951" to get your promotional price.
This item is not returnable.
View return policy
Description
Commonly used buffering agent
HEPES is commonly used as a buffering agent.Specifications
0.01 max. (0.1M solution) at 280nm | |
7365-45-9 | |
5.0 to 6.5 | |
100 g | |
DNase-, RNase- and Protease-Free | |
Pass Test | |
MFCD00006158 | |
Protease free | |
N-(2-Hydroxyethyl)piperazine-N'-2-ethanesulfonic Acid | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid | |
23831 | |
≥99% |
HEPES | |
White | |
Powder or Solid | |
DNase free | |
≥99 % | |
C8H18N2O4S | |
Glass Bottle | |
15, 4689 | |
Soluble in water | |
OCCN1CCN(CCS(O)(=O)=O)CC1 | |
238.30 | |
CHEBI:42334 | |
Molecular Biology |
Safety and Handling
Product Identifier
- HEPES
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction