missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
Commonly used buffering agent.
Brand: Fisher BioReagents BP310-5
749.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Commonly used buffering agent
HEPES is commonly used as a buffering agent.Specifications
0.01 max. (0.1M solution) at 280nm | |
7365-45-9 | |
5.0 to 6.5 | |
5 kg | |
N-2-Hydroxyethylpiperazine-N-2-Ethane Sulfonic Acid | |
≥99 % | |
C8H18N2O4S | |
Poly Pail | |
15, 4689 | |
Soluble in water | |
OCCN1CCN(CCS(O)(=O)=O)CC1 | |
238.30 | |
CHEBI:42334 | |
Molecular Biology |
HEPES | |
White | |
Crystals | |
DNase free | |
DNase-, RNase- and Protease-Free | |
Pass Test | |
MFCD00006158 | |
Protease free | |
N-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid | |
23831 | |
≥99% |
Safety and Handling
Product Identifier
- HEPES