missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES, free acid, >98%, MP Biomedicals™
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
500 g
1 kg
5 kg
Specifications
Specifications
Color | White |
Assay Percent Range | >98% |
Quantity | 100 g |
IUPAC Name | 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid |
Formula Weight | 238.3 |
Physical Form | Powder |
Chemical Name or Material | HEPES |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction